Butyl heptafluorobutyrate 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Butyl perfluorobutyrate
PC0960-10G
PC0960-50G
PC0960-100G
APOSPC0960-10GEA
992
CZK
APOSPC0960-10G
APOSPC0960-50G
APOSPC0960-100G
Butyl heptafluorobutyrate 98%
Butyl heptafluorobutyrate
Vzorec:
C₈H₉F₇O₂ MW: 270,15 g/mol Bod varu: 131…133 °C/740mmHg Hustota: 1,296 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00039240 CAS číslo: 1559-07-5 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...