Tisk…

N-Boc-3-trifluoromethyl-L-phenylalanine 97%

Dodavatel: Apollo Scientific

Synonyms: Boc-Phe(3-CF3)-OH, (S)-2-((tert-Butoxycarbonyl)amino)-3-(3-(trifluoromethyl)phenyl)propanoic acid

PC0938-10G PC0938-1G PC0938-5G
APOSPC0938-10GEA 25890 CZK
APOSPC0938-10G APOSPC0938-1G APOSPC0938-5G
N-Boc-3-trifluoromethyl-L-phenylalanine 97%
N-Boc-3-trifluoromethyl-L-phenylalanine
Vzorec: C₁₅H₁₈F₃NO₄
MW: 333,307 g/mol
Bod tání: 135…138 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00672524
CAS číslo: 142995-31-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR