Tisk…

3-(Trifluoromethoxy)benzenesulphonyl chloride 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 3-(Trifluoromethoxy)benzene-1-sulphonyl chloride

PC0934-100G PC0934-25G PC0934-5G
APOSPC0934-100GEA 6350 CZK
APOSPC0934-100G APOSPC0934-25G APOSPC0934-5G
3-(Trifluoromethoxy)benzenesulphonyl chloride 97%
3-(Trifluoromethoxy)benzenesulphonyl chloride
Vzorec: C₇H₄ClF₃O₃S
MW: 260.62
Bod varu: 229…230 °C
Hustota: 1.53 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD01091016
CAS číslo: 220227-84-9
UN: 3265
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR