Tisk…

3,5-Difluoro-2-nitrobenzoic acid

Dodavatel: Apollo Scientific
Varování

PC0922-1G PC0922-25G PC0922-5G PC0922-100G
APOSPC0922-1GEA 910 CZK
APOSPC0922-1G APOSPC0922-25G APOSPC0922-5G APOSPC0922-100G
3,5-Difluoro-2-nitrobenzoic acid
3,5-Difluoro-2-nitrobenzoic acid
Vzorec: C₇H₃F₂NO₄
MW: 203,0998 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04035649
CAS číslo: 331765-71-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR