2-Fluoro-6-methoxybenzyl alcohol
Dodavatel: Apollo Scientific
|
Varování
|
PC0894-1G
PC0894-25G
PC0894-5G
APOSPC0894-1GEA
622
CZK
APOSPC0894-1G
APOSPC0894-25G
APOSPC0894-5G
2-Fluoro-6-methoxybenzyl alcohol
2-Fluoro-6-methoxybenzyl alcohol
Vzorec:
C₈H₉FO₂ Bod tání: 35…39 °C Storage Temperature: Ambient |
MDL Number:
MFCD00671766 CAS číslo: 253668-46-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...