Tisk…

3-Fluoro-2-methylphenylacetonitrile 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-(3-Fluoro-2-methylphenyl)acetonitrile

PC0888-1G PC0888-5G PC0888-25G
APOSPC0888-1GEA 1040 CZK
APOSPC0888-1G APOSPC0888-5G APOSPC0888-25G
3-Fluoro-2-methylphenylacetonitrile 98%
3-Fluoro-2-methylphenylacetonitrile
Vzorec: C₉H₈FN
Storage Temperature: Ambient
MDL Number: MFCD03788544
CAS číslo: 500912-15-2
UN: 3439
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR