Tisk…

3-Fluoro-2-methylbenzyl bromide 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC0887-1G PC0887-5G PC0887-10G
APOSPC0887-1GEA 1820 CZK
APOSPC0887-1G APOSPC0887-5G APOSPC0887-10G
3-Fluoro-2-methylbenzyl bromide 98%
3-Fluoro-2-methylbenzyl bromide
Vzorec: C₈H₈BrF
MW: 203.05
Storage Temperature: Ambient
MDL Number: MFCD03788543
CAS číslo: 500912-14-1
UN: 3265
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR