3-Fluoro-2-methylbenzoyl chloride 98%
Dodavatel: Apollo Scientific
PC0884-5G
PC0884-1G
APOSPC0884-5GEA
4240
CZK
APOSPC0884-5G
APOSPC0884-1G
3-Fluoro-2-methylbenzoyl chloride 98%
3-Fluoro-2-methylbenzoyl chloride
Vzorec:
C₈H₆ClFO Storage Temperature: Ambient |
MDL Number:
MFCD01631251 CAS číslo: 168080-76-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...