3-Fluoro-o-toluonitrile 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC0883-25G
PC0883-50G
PC0883-5G
APOSPC0883-25GEA
1820
CZK
APOSPC0883-25G
APOSPC0883-50G
APOSPC0883-5G
3-Fluoro-o-toluonitrile 98%
3-Fluoro-o-toluonitrile
Vzorec:
C₈H₆FN Storage Temperature: Ambient |
MDL Number:
MFCD03788541 CAS číslo: 185147-06-2 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...