3-Fluoro-o-toluic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC0882-100G
PC0882-25G
PC0882-5G
APOSPC0882-100GEA
4800
CZK
APOSPC0882-100G
APOSPC0882-25G
APOSPC0882-5G
3-Fluoro-o-toluic acid 98%
3-Fluoro-o-toluic acid
Vzorec:
C₈H₇FO₂ MW: 154,14 g/mol Bod tání: 157…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075246 CAS číslo: 699-90-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...