2,4,6-Trifluorobenzenesulphonyl chloride 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2,4,6-Trifluorobenzene-1-sulphonyl chloride
PC0878-1G
PC0878-5G
APOSPC0878-1GEA
827
CZK
APOSPC0878-1G
APOSPC0878-5G
2,4,6-Trifluorobenzenesulphonyl chloride 95%
2,4,6-Trifluorobenzenesulphonyl chloride
Vzorec:
C₆H₂ClF₃O₂S MW: 230,59 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01091015 CAS číslo: 220239-64-5 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...