Tisk…

Methyl-2-fluoro-6-methoxybenzoate

Dodavatel: Apollo Scientific
Varování

Synonyms: Methyl 2-fluoro-6-methoxybenzoate, 2-Fluoro-6-methoxybenzoic acid methyl ester

PC0870-1G PC0870-5G PC0870-25G
APOSPC0870-1GEA 622 CZK
APOSPC0870-1G APOSPC0870-5G APOSPC0870-25G
Methyl-2-fluoro-6-methoxybenzoate
Methyl-2-fluoro-6-methoxybenzoate
Vzorec: C₉H₉FO₃
MW: 184,17 g/mol
Bod vzplanutí: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD03788537
CAS číslo: 178747-79-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR