Tisk…

Pentafluorophenylboronic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2,3,4,5,6-Pentafluorobenzeneboronic acid, Dihydroxy(pentafluorophenyl)borane, (Pentafluorophenyl)metaboric acid, Pentafluorobenzeneboronic acid

PC0869-5G PC0869-10G PC0869-1G
APOSPC0869-5GEA 1780 CZK
APOSPC0869-5G APOSPC0869-10G APOSPC0869-1G
Pentafluorophenylboronic acid 97%
Pentafluorophenylboronic acid
Vzorec: C₆H₂BF₅O₂
Bod tání: 250…255 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01074663
CAS číslo: 1582-24-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR