6-Fluoro-m-toluic acid
Dodavatel: Apollo Scientific
|
Varování
|
PC0854-25G
PC0854-5G
APOSPC0854-25GEA
1450
CZK
APOSPC0854-25G
APOSPC0854-5G
6-Fluoro-m-toluic acid
6-Fluoro-m-toluic acid
Vzorec:
C₈H₇FO₂ MW: 154,14 g/mol Bod tání: 160…162 °C Storage Temperature: Ambient |
MDL Number:
MFCD00092819 CAS číslo: 321-12-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...