Tisk…

α-Chloro-3,5-difluorotoulene 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1-(Chloromethyl)-3,5-difluorobenzene, 3,5-Difluorobenzyl chloride

PC0852-100G PC0852-25G PC0852-5G
APOSPC0852-100GEA 6450 CZK
APOSPC0852-100G APOSPC0852-25G APOSPC0852-5G
α-Chloro-3,5-difluorotoulene 98%
α-Chloro-3,5-difluorotoulene
Vzorec: C₇H₅ClF₂
Bod varu: 82 °C/22mmHg
Storage Temperature: Ambient
MDL Number: MFCD01090990
CAS číslo: 220141-71-9
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR