Tisk…

5-Bromo-3-(trifluoromethoxy)salicylaldehyde

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Bromo-2-hydroxy-3-(trifluoromethoxy)benzaldehyde

PC0844-1G PC0844-250MG
APOSPC0844-1GEA 11710 CZK
APOSPC0844-1G APOSPC0844-250MG
5-Bromo-3-(trifluoromethoxy)salicylaldehyde
5-Bromo-3-(trifluoromethoxy)salicylaldehyde
Vzorec: C₈H₄BrF₃O₃
Storage Temperature: Ambient
MDL Number: MFCD03788532
CAS číslo: 497959-32-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR