5-Bromo-3-(trifluoromethyl)salicylaldehyde 99%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Bromo-2-hydroxy-3-(trifluoromethyl)benzaldehyde
PC0843-10G
PC0843-1G
PC0843-5G
APOSPC0843-10GEA
28040
CZK
APOSPC0843-10G
APOSPC0843-1G
APOSPC0843-5G
5-Bromo-3-(trifluoromethyl)salicylaldehyde 99%
5-Bromo-3-(trifluoromethyl)salicylaldehyde
Vzorec:
C₈H₄BrF₃O₂ MW: 269,200 g/mol Bod tání: 90…92 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03788531 CAS číslo: 251300-30-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...