3-(Trifluoromethoxy)salicylaldehyde 99%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Hydroxy-3-(trifluoromethoxy)benzaldehyde
PC0839-1G
PC0839-250MG
APOSPC0839-1GEA
2570
CZK
APOSPC0839-1G
APOSPC0839-250MG
3-(Trifluoromethoxy)salicylaldehyde 99%
3-(Trifluoromethoxy)salicylaldehyde
Vzorec:
C₈H₅F₃O₃ MW: 206,12 g/mol Bod tání: 56…60 °C Storage Temperature: Ambient |
MDL Number:
MFCD03788527 CAS číslo: 497959-31-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...