Tisk…

4-(3-(Trifluoromethyl)-1H-pyrazol-1-yl)aniline 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-(4-Aminophenyl)-3-(trifluoromethyl)pyrazole

PC0831-1G PC0831-5G PC0831-10G
APOSPC0831-1GEA 3740 CZK
APOSPC0831-1G APOSPC0831-5G APOSPC0831-10G
4-(3-(Trifluoromethyl)-1H-pyrazol-1-yl)aniline 97%
4-(3-(Trifluoromethyl)-1H-pyrazol-1-yl)aniline
MW: 227,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00115025
CAS číslo: 231953-33-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR