Tisk…

1-(Methylsulphonyl)-4-(trifluoromethoxy)benzene 98%

Dodavatel: Apollo Scientific

PC0819-10G PC0819-5G
APOSPC0819-10GEA 9760 CZK
APOSPC0819-10G APOSPC0819-5G
1-(Methylsulphonyl)-4-(trifluoromethoxy)benzene 98%
1-(Methylsulphonyl)-4-(trifluoromethoxy)benzene
Vzorec: C₈H₇F₃O₃S
Bod tání: 64…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00792426
CAS číslo: 87750-51-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR