Tisk…

2,2-Difluoroethylamine 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC0806-5G PC0806-25G
APOSPC0806-5GEA 662 CZK
APOSPC0806-5G APOSPC0806-25G
2,2-Difluoroethylamine 97%
2,2-Difluoroethylamine
Vzorec: C₂H₅F₂N
Bod varu: 68…69 °C
Hustota: 1,15 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00798137
CAS číslo: 430-67-1
UN: 2735
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR