Tisk…

4-(Pentafluorothio)aniline 98+%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-Aminophenylsulphur pentafluoride

PC0780-1G PC0780-5G PC0780-25G
APOSPC0780-1GEA 1450 CZK
APOSPC0780-1G APOSPC0780-5G APOSPC0780-25G
4-(Pentafluorothio)aniline 98+%
4-(Pentafluorothio)aniline
Vzorec: C₆H₆F₅NS
MW: 219,18 g/mol
Bod tání: 64…67 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01076203
CAS číslo: 2993-24-0
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR