Tisk…

Pyridinium fluorochromate 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC0727-25G PC0727-5G
APOSPC0727-25GEA 5200 CZK
APOSPC0727-25G APOSPC0727-5G
Pyridinium fluorochromate 98%
Pyridinium fluorochromate
Vzorec: C₅H₆CrFNO₃
MW: 199,1 g/mol
Bod tání: 165 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00151617
CAS číslo: 83042-08-4
UN: 1479
ADR: 5.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR