Tisk…

4-(Difluoromethoxy)benzoyl chloride 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC0681-1G PC0681-5G
APOSPC0681-1GEA 3540 CZK
APOSPC0681-1G APOSPC0681-5G
4-(Difluoromethoxy)benzoyl chloride 97%
4-(Difluoromethoxy)benzoyl chloride
Vzorec: C₈H₅ClF₂O₂
Bod varu: 116…118 °C/15mmHg
Storage Temperature: Ambient
MDL Number: MFCD00833410
CAS číslo: 57320-63-5
UN: 3265
ADR: 8,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR