Tisk…

(S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: (S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethyl alcohol, (S)-(-)-3,5-Bis(trifluoromethyl)-alpha-methylbenzyl alcohol, (1S)-(-)-1-[3,5-Bis(trifluoromethyl)phenyl]ethan-1-ol, (S)-1-(3,5-Bis(trifluoromethyl)phenyl)ethanol, (S)-3,5-Bis(trifluoromethyl)-alpha-methylbenzyl alcohol

PC0581-1G PC0581-25G PC0581-5G
APOSPC0581-1GEA 622 CZK
APOSPC0581-1G APOSPC0581-25G APOSPC0581-5G
(S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol 98%
(S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol
Vzorec: C₁₀H₈F₆O
MW: 258,16 g/mol
Bod varu: 83 °C/11mmHg
Storage Temperature: Ambient
MDL Number: MFCD03093011
CAS číslo: 225920-05-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR