Tisk…

3,3-Dichloro-1,1,1-trifluoroacetone hydrate

Dodavatel: Apollo Scientific
Varování

PC0565-100G PC0565-25G
APOSPC0565-100GEA 18680 CZK
APOSPC0565-100G APOSPC0565-25G
3,3-Dichloro-1,1,1-trifluoroacetone hydrate
3,3-Dichloro-1,1,1-trifluoroacetone hydrate
Vzorec: C₃H₃Cl₂F₃O₂
Bod varu: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD04038304
CAS číslo: 1049731-87-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR