Tisk…

3,4,5-Trifluoro-DL-phenylalanine

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Amino-3-(3,4,5-trifluorophenyl)propanoic acid, H-DL-Phe(3,4,5-TriF)-OH

PC0531-1G PC0531-5G PC0531-10G
APOSPC0531-1GEA 1660 CZK
APOSPC0531-1G APOSPC0531-5G APOSPC0531-10G
3,4,5-Trifluoro-DL-phenylalanine
3,4,5-Trifluoro-DL-phenylalanine
Vzorec: C₉H₈F₃NO₂
MW: 219,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631414
CAS číslo: 261952-26-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR