Tisk…

Bis(hexafluoroisopropyl)itaconate 95%

Dodavatel: Apollo Scientific
Varování

PC0528-25G PC0528-5G
APOSPC0528-25GEA 12440 CZK
APOSPC0528-25G APOSPC0528-5G
Bis(hexafluoroisopropyl)itaconate 95%
Bis(hexafluoroisopropyl)itaconate
Vzorec: C₁₁H₆F₁₂O₄
Storage Temperature: Refrigerator
MDL Number: MFCD00042168
CAS číslo: 98452-82-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR