3-(Trifluoromethoxy)anisole 98%
Dodavatel: Apollo Scientific
Synonyms:
1-Methoxy-3-(trifluoromethoxy)benzene
PC0516-25G
PC0516-5G
PC0516-10G
APOSPC0516-25GEA
3710
CZK
APOSPC0516-25G
APOSPC0516-5G
APOSPC0516-10G
3-(Trifluoromethoxy)anisole 98%
3-(Trifluoromethoxy)anisole
Vzorec:
C₈H₇F₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD01631504 CAS číslo: 142738-94-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...