2,3,6-Trifluoroanisole 98%
Dodavatel: Apollo Scientific
PC0510-1G
PC0510-250MG
APOSPC0510-1GEA
705
CZK
APOSPC0510-1G
APOSPC0510-250MG
2,3,6-Trifluoroanisole 98%
2,3,6-Trifluoroanisole
Vzorec:
C₇H₅F₃O MW: 162,11 g/mol Bod varu: 146 °C Bod vzplanutí: 45 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631501 CAS číslo: 4920-34-7 UN: 3271 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...