Tisk…

2,3,4-Trifluoroanisole 98%

Dodavatel: Apollo Scientific

Synonyms: 1,2,3-Trifluoro-4-methoxybenzene

PC0507-25G PC0507-5G PC0507-100G
APOSPC0507-25GEA 6950 CZK
APOSPC0507-25G APOSPC0507-5G APOSPC0507-100G
2,3,4-Trifluoroanisole 98%
2,3,4-Trifluoroanisole
Vzorec: C₇H₅F₃O
MW: 162,11 g/mol
Bod varu: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00042396
CAS číslo: 203245-16-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR