Tisk…

3-Methyl-5-(trifluoromethyl)benzyl bromide

Dodavatel: Apollo Scientific
Nebezpečí

PC0483-1G PC0483-5G
APOSPC0483-1GEA 9890 CZK
APOSPC0483-1G APOSPC0483-5G
3-Methyl-5-(trifluoromethyl)benzyl bromide
3-Methyl-5-(trifluoromethyl)benzyl bromide
Vzorec: C₉H₈BrF₃
Storage Temperature: Ambient
MDL Number: MFCD01631610
CAS číslo: 116070-36-1
UN: 3265
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR