Tisk…

8-Fluoro-4-hydroxyquinoline-3-carboxylic acid

Dodavatel: Apollo Scientific
Varování

PC0482-1G PC0482-250MG
APOSPC0482-1GEA 6150 CZK
APOSPC0482-1G APOSPC0482-250MG
8-Fluoro-4-hydroxyquinoline-3-carboxylic acid
8-Fluoro-4-hydroxyquinoline-3-carboxylic acid
Vzorec: C₁₀H₆FNO₃
Storage Temperature: Ambient
MDL Number: MFCD00218201
CAS číslo: 63010-70-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR