Tisk…

2-Methyl-5-(trifluoromethyl)benzyl bromide 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC0480-1G PC0480-5G PC0480-10G
APOSPC0480-1GEA 992 CZK
APOSPC0480-1G APOSPC0480-5G APOSPC0480-10G
2-Methyl-5-(trifluoromethyl)benzyl bromide 98%
2-Methyl-5-(trifluoromethyl)benzyl bromide
Vzorec: C₉H₈BrF₃
MW: 253.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631609
CAS číslo: 261952-17-4
UN: 3265
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR