4-Methyl-3-(trifluoromethyl)benzonitrile 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC0460-25G
PC0460-1G
PC0460-5G
APOSPC0460-25GEA
3310
CZK
APOSPC0460-25G
APOSPC0460-1G
APOSPC0460-5G
4-Methyl-3-(trifluoromethyl)benzonitrile 98%
4-Methyl-3-(trifluoromethyl)benzonitrile
Vzorec:
C₉H₆F₃N Bod tání: 60…62 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631597 CAS číslo: 261952-06-1 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...