3-Methyl-5-(trifluoromethyl)benzonitrile 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC0456-5G
PC0456-10G
PC0456-1G
APOSPC0456-5GEA
7670
CZK
APOSPC0456-5G
APOSPC0456-10G
APOSPC0456-1G
3-Methyl-5-(trifluoromethyl)benzonitrile 97%
3-Methyl-5-(trifluoromethyl)benzonitrile
Vzorec:
C₉H₆F₃N Storage Temperature: Ambient |
MDL Number:
MFCD01631595 CAS číslo: 261952-04-9 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...