Tisk…

2-Methyl-4-(trifluoromethoxy)benzyl bromide 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC0431-1G PC0431-5G
APOSPC0431-1GEA 4070 CZK
APOSPC0431-1G APOSPC0431-5G
2-Methyl-4-(trifluoromethoxy)benzyl bromide 97%
2-Methyl-4-(trifluoromethoxy)benzyl bromide
Vzorec: C₉H₈BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD01631547
CAS číslo: 261951-95-5
UN: 1760
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR