5-Fluoro-2-(trifluoromethyl)cinnamic acid
Dodavatel: Apollo Scientific
PC0423-10G
PC0423-5G
APOSPC0423-10GEA
5030
CZK
APOSPC0423-10G
APOSPC0423-5G
5-Fluoro-2-(trifluoromethyl)cinnamic acid
5-Fluoro-2-(trifluoromethyl)cinnamic acid
Vzorec:
C₁₀H₆F₄O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00236299 CAS číslo: 231291-18-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...