Tisk…

4-Bromo-2-(trifluoromethyl)anisole 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Bromo-2-methoxybenzotrifluoride, 4-Methoxy-3-(trifluoromethyl)bromobenzene

PC0413-100G PC0413-25G PC0413-5G
APOSPC0413-100GEA 4930 CZK
APOSPC0413-100G APOSPC0413-25G APOSPC0413-5G
4-Bromo-2-(trifluoromethyl)anisole 98%
4-Bromo-2-(trifluoromethyl)anisole
Vzorec: C₈H₆BrF₃O
MW: 255,03 g/mol
Bod varu: 120 °C/30mmHg
Storage Temperature: Ambient
MDL Number: MFCD01631579
CAS číslo: 1514-11-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR