Tisk…

2-Fluoro-3-(trifluoromethyl)anisole 98%

Dodavatel: Apollo Scientific

PC0380-5G PC0380-1G
APOSPC0380-5GEA 3740 CZK
APOSPC0380-5G APOSPC0380-1G
2-Fluoro-3-(trifluoromethyl)anisole 98%
2-Fluoro-3-(trifluoromethyl)anisole
Vzorec: C₈H₆F₄O
MW: 194,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631563
CAS číslo: 151868-17-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR