Tisk…

2-Fluoro-4-(trifluoromethyl)aniline

Dodavatel: Apollo Scientific
Varování

PC0379-100G PC0379-25G PC0379-5G
APOSPC0379-100GEA 14680 CZK
APOSPC0379-100G APOSPC0379-25G APOSPC0379-5G
2-Fluoro-4-(trifluoromethyl)aniline
2-Fluoro-4-(trifluoromethyl)aniline
Vzorec: C₇H₅F₄N
Bod varu: 55 °C/0,3mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD01631629
CAS číslo: 69409-98-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR