Tisk…

3-Fluoro-4-methyl-DL-phenylglycine

Dodavatel: Apollo Scientific
Varování

PC0373-1G PC0373-5G PC0373-10G
APOSPC0373-1GEA 3310 CZK
APOSPC0373-1G APOSPC0373-5G APOSPC0373-10G
3-Fluoro-4-methyl-DL-phenylglycine
3-Fluoro-4-methyl-DL-phenylglycine
Vzorec: C₉H₁₀FNO₂
Storage Temperature: Ambient
MDL Number: MFCD06247587
CAS číslo: 261951-76-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR