5-Fluoro-2-methylphenylacetonitrile 97%
Dodavatel: Apollo Scientific
Synonyms:
2-(5-Fluoro-2-methylphenyl)acetonitrile
PC0369-5G
PC0369-1G
APOSPC0369-5GEA
2480
CZK
APOSPC0369-5G
APOSPC0369-1G
5-Fluoro-2-methylphenylacetonitrile 97%
5-Fluoro-2-methylphenylacetonitrile
Vzorec:
C9H8FN MW: 149,1700 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01631529 CAS číslo: 80141-97-5 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...