3-Fluoro-4-methylphenylacetonitrile 98%
Dodavatel: Apollo Scientific
PC0368-5G
PC0368-1G
PC0368-25G
APOSPC0368-5GEA
2900
CZK
APOSPC0368-5G
APOSPC0368-1G
APOSPC0368-25G
3-Fluoro-4-methylphenylacetonitrile 98%
3-Fluoro-4-methylphenylacetonitrile
Vzorec:
C₉H₈FN Storage Temperature: Ambient |
MDL Number:
MFCD01631535 CAS číslo: 261951-73-9 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...