Tisk…

5-Fluoro-2-methylbenzylamine 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: (5-Fluoro-2-methylphenyl)methanamine

PC0359-1G PC0359-25G PC0359-5G
APOSPC0359-1GEA 827 CZK
APOSPC0359-1G APOSPC0359-25G APOSPC0359-5G
5-Fluoro-2-methylbenzylamine 98%
5-Fluoro-2-methylbenzylamine
Vzorec: C₈H₁₀FN
MW: 139,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631526
CAS číslo: 261951-69-3
UN: 2735
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR