3-Fluoro-4-methylbenzylamine 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC0356-5G
PC0356-1G
APOSPC0356-5GEA
2860
CZK
APOSPC0356-5G
APOSPC0356-1G
3-Fluoro-4-methylbenzylamine 97%
3-Fluoro-4-methylbenzylamine
Vzorec:
C₈H₁₀FN MW: 139,17 g/mol Hustota: 1,05 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD01631531 CAS číslo: 261951-67-1 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...