Tisk…

3-Fluoro-4-methylbenzyl alcohol

Dodavatel: Apollo Scientific
Varování

Synonyms: (3-Fluoro-4-methylphenyl)methanol

PC0351-10G PC0351-1G PC0351-5G PC0351-25G
APOSPC0351-10GEA 4530 CZK
APOSPC0351-10G APOSPC0351-1G APOSPC0351-5G APOSPC0351-25G
3-Fluoro-4-methylbenzyl alcohol
3-Fluoro-4-methylbenzyl alcohol
Vzorec: C₈H₉FO
MW: 140,16 g/mol
Bod tání: 33…36 °C
Storage Temperature: Ambient
MDL Number: MFCD01631533
CAS číslo: 192702-79-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR