Tisk…

3-Fluoro-4-(trifluoromethyl)cinnamic acid

Dodavatel: Apollo Scientific
Varování

PC0347-1G PC0347-5G PC0347-25G PC0347-100G
APOSPC0347-1GEA 622 CZK
APOSPC0347-1G APOSPC0347-5G APOSPC0347-25G APOSPC0347-100G
3-Fluoro-4-(trifluoromethyl)cinnamic acid
3-Fluoro-4-(trifluoromethyl)cinnamic acid
Vzorec: C₁₀H₆F₄O₂
Storage Temperature: Ambient
MDL Number: MFCD00236295
CAS číslo: 231291-13-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR