2,3,5,6-Tetrafluoro-4-hydroxybenzoic acid
Dodavatel: Apollo Scientific
|
Varování
|
PC0330-5G
APOSPC0330-5GEA
3020
CZK
APOSPC0330-5G
2,3,5,6-Tetrafluoro-4-hydroxybenzoic acid
2,3,5,6-Tetrafluoro-4-hydroxybenzoic acid
Vzorec:
C₇H₂F₄O₃ MW: 210,08 g/mol Bod tání: 148…150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00129952 CAS číslo: 652-34-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...