1-(4-Fluoro-α-phenylbenzyl)piperazine
Dodavatel: Apollo Scientific
PC0329-1G
PC0329-5G
APOSPC0329-1GEA
6220
CZK
APOSPC0329-1G
APOSPC0329-5G
1-(4-Fluoro-α-phenylbenzyl)piperazine
1-(4-Fluoro-alpha-phenylbenzyl)piperazine
Vzorec:
C₁₇H₁₉FN₂ Storage Temperature: Ambient |
MDL Number:
MFCD00426601 CAS číslo: 27064-89-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...